Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azide-PEG5-tos | 236754-49-7 | MFCD28505520 | 98.5% | 417.48 | C17H27N3O7S | 5 G
SDP

Catalog No. 5000202627
Encompass_Preferred

Azide-PEG5-Tos is a cleavable 5-unit polyethylene glycol (PEG) linker bearing an azide functional group and a tosyl leaving group, designed for click-chemistry conjugations and the synthesis of antibody-drug conjugates and PROTAC linkers.

  • cleavable 5-unit peg linker suitable for ADC and PROTAC synthesis.
  • azide functional group for CuAAC and SPAAC click reactions.
  • high purity suitable for research applications.
  • appearance: liquid, colorless to light yellow.
  • molecular formula c17h27n3o7s and molecular weight 417.48 g/mol.
  • storage: pure form at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-02627 Supplier Medchemexpress LLC Supplier No. HY1403525G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azide-PEG5-Tos is a cleavable 5-unit polyethylene glycol (PEG) linker bearing an azide functional group and a tosyl leaving group, designed for click-chemistry conjugations and the synthesis of antibody-drug conjugates and PROTAC linkers.

  • cleavable 5-unit peg linker suitable for ADC and PROTAC synthesis.
  • azide functional group for CuAAC and SPAAC click reactions.
  • high purity suitable for research applications.
  • appearance: liquid, colorless to light yellow.
  • molecular formula c17h27n3o7s and molecular weight 417.48 g/mol.
  • storage: pure form at -20°C (3 years) or 4°C (2 years); in solvent at -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.