Learn More
Medchemexpress LLC Azide-PEG8-alcohol | 352439-36-2 | 97.0% | C16H33N3O8 | 50 MG
SDP
Catalog No. 5000355500
Encompass_Preferred
Azide-PEG8-alcohol is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. It functions as a versatile click chemistry reagent.
  • Contains an azide group for click chemistry
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Useful for PROTAC synthesis
Catalog No. 50-003-55500 Supplier Medchemexpress LLC Supplier No. HY14079950MG
Azide-PEG8-alcohol is a PEG-based PROTAC linker primarily utilized in the synthesis of PROTACs. It functions as a versatile click chemistry reagent.
  • Contains an azide group for click chemistry
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Useful for PROTAC synthesis