Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-AHPC-PEG3 (VHL ligand-linker) pyrrolidine-2-carboxamide | 1797406-80-4 | MFCD31807299 | ≥95.0% | 645.77 g/mol | C30H43N7O7S | 10 MG
SDP

Catalog No. 5000247669
Encompass_Preferred

(S,R,S)-AHPC-PEG3-N3 is an E3 ligase ligand-linker conjugate that couples an (S,R,S)-AHPC VHL-binding motif to a three-unit PEG linker terminated with an azide. It is intended as a click-chemistry reagent for PROTAC synthesis and bioconjugation, enabling covalent linkage to alkyne-bearing partners.

  • Molecular weight 645.77 g/mol.
  • Chemical formula C30H43N7O7S.
  • CAS number 1797406-80-4.
  • Purity ≥95%.
  • Suitable for CuAAC and SPAAC click reactions.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Soluble in ethanol, water, and DMSO at concentrations commonly used for bioconjugation.

Catalog No. 50-002-47669 Supplier Medchemexpress LLC Supplier No. HY10359810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(S,R,S)-AHPC-PEG3-N3 is an E3 ligase ligand-linker conjugate that couples an (S,R,S)-AHPC VHL-binding motif to a three-unit PEG linker terminated with an azide. It is intended as a click-chemistry reagent for PROTAC synthesis and bioconjugation, enabling covalent linkage to alkyne-bearing partners.

  • Molecular weight 645.77 g/mol.
  • Chemical formula C30H43N7O7S.
  • CAS number 1797406-80-4.
  • Purity ≥95%.
  • Suitable for CuAAC and SPAAC click reactions.
  • Storage: pure form -20°C (3 years) or 4°C (2 years); in solvent -80°C (6 months) or -20°C (1 month).
  • Soluble in ethanol, water, and DMSO at concentrations commonly used for bioconjugation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.