Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-choline bromide | 2059973-54-3 | 99.6% | 239.11 g·mol⁻¹ | C6H15BrN4O | 1 ML
SDP

Catalog No. 5000284584
Encompass_Preferred

N3-Cho bromide (azido-choline bromide) is an azide-bearing choline analogue supplied as a ready-to-use 10 mM solution in DMSO for bioorthogonal labeling and click chemistry applications. It is intended for incorporation into membrane mimetics and for use in azide-alkyne cycloaddition workflows.

  • Contains an azide functional group compatible with click chemistry reactions.
  • Supplied as a 10 mM solution in DMSO, ready for dilution or reaction.
  • High purity (listed at 99.63%), suitable for research use.
  • Soluble in DMSO and water.
  • Stable in solution when sealed at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Useful for membrane labeling and synthesis of phospholipid head-group analogues.

Catalog No. 50-002-84584 Supplier Medchemexpress LLC Supplier No. HYW588725A10MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N3-Cho bromide (azido-choline bromide) is an azide-bearing choline analogue supplied as a ready-to-use 10 mM solution in DMSO for bioorthogonal labeling and click chemistry applications. It is intended for incorporation into membrane mimetics and for use in azide-alkyne cycloaddition workflows.

  • Contains an azide functional group compatible with click chemistry reactions.
  • Supplied as a 10 mM solution in DMSO, ready for dilution or reaction.
  • High purity (listed at 99.63%), suitable for research use.
  • Soluble in DMSO and water.
  • Stable in solution when sealed at -80°C (up to 6 months) or -20°C (up to 1 month).
  • Useful for membrane labeling and synthesis of phospholipid head-group analogues.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.