Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-choline bromide | 2059973-54-3 | 99.6% | 239.11 g/mol | C6H15BrN4O | 25 MG
SDP

Catalog No. 5000284276
Encompass_Preferred

N3-Cho bromide (Azido-choline bromide) is an azide-functionalized choline analogue used as a click chemistry reagent to install or label choline head-groups in membrane lipids and related biomolecules. It is provided as a colorless to off-white solid-liquid mixture with high purity and good solubility in common solvents, intended for synthetic and labeling applications.

  • Azide-functionalized choline analogue for click chemistry labeling.
  • High purity suitable for synthetic and analytical use.
  • Soluble in water (125 mg/mL) and DMSO (100 mg/mL); ultrasonic recommended.
  • Storage recommendations for stability: solid at -20°C sealed; in solvent -80°C up to 6 months.
  • Available in small research pack sizes for laboratory experiments.

Catalog No. 50-002-84276 Supplier Medchemexpress LLC Supplier No. HYW588725A25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

N3-Cho bromide (Azido-choline bromide) is an azide-functionalized choline analogue used as a click chemistry reagent to install or label choline head-groups in membrane lipids and related biomolecules. It is provided as a colorless to off-white solid-liquid mixture with high purity and good solubility in common solvents, intended for synthetic and labeling applications.

  • Azide-functionalized choline analogue for click chemistry labeling.
  • High purity suitable for synthetic and analytical use.
  • Soluble in water (125 mg/mL) and DMSO (100 mg/mL); ultrasonic recommended.
  • Storage recommendations for stability: solid at -20°C sealed; in solvent -80°C up to 6 months.
  • Available in small research pack sizes for laboratory experiments.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.