Learn More
Cayman Chemical Azido-dPEG7-amine, Purity: ≥90%, CAS: 1333154-77-0, 10 mg
SDP
Catalog No. 502992232
Encompass_Preferred
A heterobifunctional PEGylated linker; contains a reactive NH2 end group, as well as an azide functional group for use in click chemistry reactions; has been used as a linker in the synthesis of bivalent mTOR inhibitors
Catalog No. 50-299-2232 Supplier Cayman Chemical Supplier No. 3054410 MG
A heterobifunctional PEGylated linker; contains a reactive NH2 end group, as well as an azide functional group for use in click chemistry reactions; has been used as a linker in the synthesis of bivalent mTOR inhibitors