Learn More
Abcam 3-Azido-7-hydro x ycoumarin, 10MG
Supplier:  Abcam AB27501410MG
Encompass_Preferred
MW 203.15 Da, Purity >98%. Novel fluorogenic 'click' labeling dye for conjugation to alkyne-containing biomolecules. Highly permeable to cell membrane and tissue. Utilizes the reaction of azide derivatives of biological molecules (sugars, lipids, etc.) with a fluorescent alkyne or cyclooctene derivative. Can be used to trace the linked biological compounds inside living cells. The detection agent employs a quenched azide dye that becomes highly fluorescent after the Click reaction when it forms triazoles at the site of alkyne modified biomolecules.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-113-9339