Learn More
Medchemexpress LLC Azido-mono-amide-DOTA | 1227407-76-2 | 100.0% | 486.52 g·mol⁻¹ | C19H34N8O7 | 10 MG
SDP
Catalog No. 5000283989
Encompass_Preferred
Azido-mono-amide-DOTA is an azide-functional DOTA derivative that acts as a bifunctional chelator for radiolabeling and bioconjugation. It combines a macrocyclic chelator for stable metal coordination with an azide handle for click chemistry, making it suitable for assembling radiopharmaceuticals and imaging probes in research settings.
  • Azide functionality enables copper-free or copper-catalyzed click conjugation.
  • Macrocyclic DOTA scaffold provides strong chelation of radiometals.
  • High reported purity (99.96%) suitable for research applications.
  • Compact molecular weight and defined chemical formula aid analytical characterization.
  • Commonly supplied in small milligram quantities for synthesis and labeling workflows.
  • CAS number 1227407-76-2 for unambiguous substance identification.
Catalog No. 50-002-83989 Supplier Medchemexpress LLC Supplier No. HYW53991610MG
Azido-mono-amide-DOTA is an azide-functional DOTA derivative that acts as a bifunctional chelator for radiolabeling and bioconjugation. It combines a macrocyclic chelator for stable metal coordination with an azide handle for click chemistry, making it suitable for assembling radiopharmaceuticals and imaging probes in research settings.
  • Azide functionality enables copper-free or copper-catalyzed click conjugation.
  • Macrocyclic DOTA scaffold provides strong chelation of radiometals.
  • High reported purity (99.96%) suitable for research applications.
  • Compact molecular weight and defined chemical formula aid analytical characterization.
  • Commonly supplied in small milligram quantities for synthesis and labeling workflows.
  • CAS number 1227407-76-2 for unambiguous substance identification.