Learn More
Medchemexpress LLC Azido-mono-amide-DOTA | 1227407-76-2 | 100.0% | 486.52 g/mol | C19H34N8O7 | 50 MG
SDP
Catalog No. 5000284091
Encompass_Preferred
Azido-mono-amide-DOTA is a macrocyclic DOTA derivative that functions as a bifunctional chelator for conjugation of peptides and radionuclides. It features an azido handle for click-chemistry attachment and is intended for use in tumor pre-targeting and radiolabeling workflows.
  • Bifunctional DOTA chelator for stable metal coordination.
  • Azido functional group enables efficient click chemistry conjugation to biomolecules.
  • High reported purity suitable for research and preclinical studies.
  • Available in small milligram quantities for method development and labeling.
  • Molecular weight 486.52, formula C19H34N8O7, CAS 1227407-76-2.
Catalog No. 50-002-84091 Supplier Medchemexpress LLC Supplier No. HYW53991650MG
Azido-mono-amide-DOTA is a macrocyclic DOTA derivative that functions as a bifunctional chelator for conjugation of peptides and radionuclides. It features an azido handle for click-chemistry attachment and is intended for use in tumor pre-targeting and radiolabeling workflows.
  • Bifunctional DOTA chelator for stable metal coordination.
  • Azido functional group enables efficient click chemistry conjugation to biomolecules.
  • High reported purity suitable for research and preclinical studies.
  • Available in small milligram quantities for method development and labeling.
  • Molecular weight 486.52, formula C19H34N8O7, CAS 1227407-76-2.