Learn More
Medchemexpress LLC Azido myristic acid | 80667-36-3 | MFCD00911365 | 98.4% | 241.33 g/mol | C12H23N3O2 | 25 MG
SDP
Catalog No. 5000202248
Encompass_Preferred
Azido myristic acid (12-azidododecanoic acid) is an azide-containing fatty acid reagent used for click-chemistry labeling and detection of protein myristoylation. It reacts with alkyne-containing probes via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and by strain-promoted azide-alkyne cycloaddition (SPAAC) with DBCO/BCN reagents, enabling metabolic labeling and enrichment of myristoylated proteins.
  • Azide functional group for click chemistry applications.
  • Suitable for metabolic labeling of myristoylated proteins.
  • Reacts in CuAAC and SPAAC conjugation reactions.
  • Supplied as a solid-liquid mixture with high purity.
  • Stable when stored under recommended conditions (see datasheet for details).
Catalog No. 50-002-02248 Supplier Medchemexpress LLC Supplier No. HY15185525MG
Azido myristic acid (12-azidododecanoic acid) is an azide-containing fatty acid reagent used for click-chemistry labeling and detection of protein myristoylation. It reacts with alkyne-containing probes via copper-catalyzed azide-alkyne cycloaddition (CuAAC) and by strain-promoted azide-alkyne cycloaddition (SPAAC) with DBCO/BCN reagents, enabling metabolic labeling and enrichment of myristoylated proteins.
  • Azide functional group for click chemistry applications.
  • Suitable for metabolic labeling of myristoylated proteins.
  • Reacts in CuAAC and SPAAC conjugation reactions.
  • Supplied as a solid-liquid mixture with high purity.
  • Stable when stored under recommended conditions (see datasheet for details).