Learn More
Medchemexpress LLC Azido-PEG(4)-Val-Cit-PAB-PNP | 1869126-60-2 | C36H51N9O13 | 50 MG
SDP
Catalog No. 5000374123
Encompass_Preferred
Azido-PEG(4)-Val-Cit-PAB-PNP is a cleavable peptide ADC linker. It functions as a click chemistry reagent, featuring an azide group. This reagent is designed to engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups, and can also participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing DBCO or BCN groups.
  • Cleavable peptide ADC linker
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Can participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
Catalog No. 50-003-74123 Supplier Medchemexpress LLC Supplier No. HY15172650MG
Azido-PEG(4)-Val-Cit-PAB-PNP is a cleavable peptide ADC linker. It functions as a click chemistry reagent, featuring an azide group. This reagent is designed to engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules containing alkyne groups, and can also participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with molecules possessing DBCO or BCN groups.
  • Cleavable peptide ADC linker
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Can participate in ring strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups