Learn More
Medchemexpress LLC Azido-PEG1 | 1517-05-1 | 100.0% | 87.08 | 100 MG
SDP
Catalog No. 5000351618
Encompass_Preferred
Azido-PEG1 is a versatile PEG-based PROTAC linker primarily utilized for synthesizing PROTACs. Its core feature is an azide group, which makes it a potent click chemistry reagent. This allows it to readily engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess alkyne groups and can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Enables the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Capable of strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Versatile reactivity
Catalog No. 50-003-51618 Supplier Medchemexpress LLC Supplier No. HY138461100MG
Azido-PEG1 is a versatile PEG-based PROTAC linker primarily utilized for synthesizing PROTACs. Its core feature is an azide group, which makes it a potent click chemistry reagent. This allows it to readily engage in copper-catalyzed azide-alkyne cycloaddition (CuAAc) with molecules that possess alkyne groups and can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Enables the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Capable of copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Capable of strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Versatile reactivity