Learn More
Medchemexpress LLC Azido-PEG1-C2-acid | 1393330-34-1 | 98.0% | 159.14 | 100 MG
SDP
Catalog No. 5000348656
Encompass_Preferred
Azido-PEG1-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent with azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Used in the synthesis of PROTACs
Catalog No. 50-003-48656 Supplier Medchemexpress LLC Supplier No. HY140009100MG
Azido-PEG1-C2-acid is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups. PROTACs leverage the intracellular ubiquitin-proteasome system to facilitate the selective degradation of target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent with azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Used in the synthesis of PROTACs