Learn More
Medchemexpress LLC Azido-PEG1-C2-acid | 1393330-34-1 | 98.0% | 159.14 | 50MG
SDP
Catalog No. 5000347994
Encompass_Preferred
Azido-PEG1-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group. This enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, and it can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for click chemistry reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-47994 Supplier Medchemexpress LLC Supplier No. HY14000950MG
Azido-PEG1-C2-acid is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group. This enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules, and it can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group for click chemistry reactions
  • Enables copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions