Learn More
Medchemexpress LLC Azido-PEG10-alcohol | 877239-09-3 | 97.0% | C20H41N3O10 | 500 MG
SDP
Catalog No. 5000352982
Encompass_Preferred
Azido-PEG10-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in various cycloaddition reactions. This product is for research use only.
  • Click chemistry reagent
  • Contains an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Used in the synthesis of PROTACs
Catalog No. 50-003-52982 Supplier Medchemexpress LLC Supplier No. HY140801500MG
Azido-PEG10-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in various cycloaddition reactions. This product is for research use only.
  • Click chemistry reagent
  • Contains an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Used in the synthesis of PROTACs