Learn More
Medchemexpress LLC Azido-PEG10-alcohol | 877239-09-3 | C20H41N3O10 | 1 G
SDP
Catalog No. 5000356482
Encompass_Preferred
Azido-PEG10-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent containing an azide group. This reagent is capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-56482 Supplier Medchemexpress LLC Supplier No. HY1408011G
Azido-PEG10-alcohol is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent containing an azide group. This reagent is capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups, as well as strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Click chemistry reagent with an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions