Learn More
Medchemexpress LLC Azido-PEG10-amine | 912849-73-1 | 97.0% | C22H46N4O10 | 100 MG
SDP
Catalog No. 5000357245
Encompass_Preferred
Azido-PEG10-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups, and also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc and SPAAC reactions
  • Enables reactions with alkyne, DBCO, or BCN groups
  • Leverages the ubiquitin-proteasome system to selectively degrade target proteins
Catalog No. 50-003-57245 Supplier Medchemexpress LLC Supplier No. HY140217100MG
Azido-PEG10-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules containing Alkyne groups, and also undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group for CuAAc and SPAAC reactions
  • Enables reactions with alkyne, DBCO, or BCN groups
  • Leverages the ubiquitin-proteasome system to selectively degrade target proteins