Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Azido-PEG10-amine | 912849-73-1 | ≥97.0% | C22H46N4O10 | 250 MG
SDP

Catalog No. 5000352862
Encompass_Preferred

Azido-PEG10-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • PROTACs degrade target proteins via the ubiquitin-proteasome system
  • Appearance is colorless to off-white

Catalog No. 50-003-52862 Supplier Medchemexpress LLC Supplier No. 250MGHY140217
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Azido-PEG10-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. This click chemistry reagent contains an Azide group, enabling participation in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.

  • PEG-based PROTAC linker for PROTAC synthesis
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups
  • PROTACs degrade target proteins via the ubiquitin-proteasome system
  • Appearance is colorless to off-white

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.