Learn More
Medchemexpress LLC Azido-PEG10-amine | 912849-73-1 | 97.0% | C22H46N4O10 | 50 MG
SDP
Catalog No. 5000353096
Encompass_Preferred
Azido-PEG10-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group, which can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-53096 Supplier Medchemexpress LLC Supplier No. HY14021750MG
Azido-PEG10-amine is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group, which can participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an Azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions