Learn More
Medchemexpress LLC Azido-PEG10-amine | 912849-73-1 | MFCD03453246 | ≥97.0% | C22H46N4O10 | 5 G
SDP
Catalog No. 5000357212
Encompass_Preferred
Azido-PEG10-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. PROTACs themselves are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilized in the synthesis of PROTACs
Catalog No. 50-003-57212 Supplier Medchemexpress LLC Supplier No. HY1402175G
Azido-PEG10-amine is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions. PROTACs themselves are designed to exploit the intracellular ubiquitin-proteasome system for the selective degradation of target proteins.
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
  • Utilized in the synthesis of PROTACs