Learn More
Medchemexpress LLC Azido-PEG10-NHS ester | 2801772-86-9 | 98.0% | C27H48N4O14 | 100 MG
SDP
Catalog No. 5000356480
Encompass_Preferred
Azido-PEG10-NHS ester is a versatile PEG-based PROTAC linker and click chemistry reagent. It contains an Azide group, facilitating copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. This compound is instrumental in the synthesis of PROTACs, which leverage the intracellular ubiquitin-proteasome system for targeted protein degradation.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Contains an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Allows reactions with alkyne-containing molecules
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with molecules containing DBCO or BCN groups
  • Aids in selective degradation of target proteins
Catalog No. 50-003-56480 Supplier Medchemexpress LLC Supplier No. HY140759100MG
Azido-PEG10-NHS ester is a versatile PEG-based PROTAC linker and click chemistry reagent. It contains an Azide group, facilitating copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups. This compound is instrumental in the synthesis of PROTACs, which leverage the intracellular ubiquitin-proteasome system for targeted protein degradation.
  • Functions as a PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Acts as a click chemistry reagent
  • Contains an Azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Allows reactions with alkyne-containing molecules
  • Supports strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with molecules containing DBCO or BCN groups
  • Aids in selective degradation of target proteins