Learn More
Medchemexpress LLC Azido-PEG11-azide | 1392284-57-9 | MFCD28385475 | 98.0% | 596.67 | C24H48N6O11 | 5 MG
SDP
Catalog No. 5000262552
Encompass_Preferred
Azido-PEG11-azide is a PEG11-based diazide linker designed for bioconjugation and linker development applications. It features terminal azide groups on an 11-unit polyethylene glycol chain, providing water solubility and flexibility for click chemistry conjugation.
  • Contains azide functional groups at both termini for azide-alkyne cycloaddition.
  • PEG11 spacer offers flexibility and improved aqueous solubility.
  • High purity suitable for research and synthetic applications.
  • Liquid form allows for easy aliquoting and handling.
  • Commonly used in bioconjugation, linker screening, and PROTAC synthesis.
Catalog No. 50-002-62552 Supplier Medchemexpress LLC Supplier No. HY1333965MG
Azido-PEG11-azide is a PEG11-based diazide linker designed for bioconjugation and linker development applications. It features terminal azide groups on an 11-unit polyethylene glycol chain, providing water solubility and flexibility for click chemistry conjugation.
  • Contains azide functional groups at both termini for azide-alkyne cycloaddition.
  • PEG11 spacer offers flexibility and improved aqueous solubility.
  • High purity suitable for research and synthetic applications.
  • Liquid form allows for easy aliquoting and handling.
  • Commonly used in bioconjugation, linker screening, and PROTAC synthesis.