Learn More
Medchemexpress LLC Azido-PEG12-amine | 2803478-25-1 | 98.0% | 614.73 g/mol | C26H54N4O12 | 50 MG
SDP
Catalog No. 5000285004
Encompass_Preferred
Azido-PEG12-amine is a PEG12 linker with an azide group at one terminus and a primary amine at the other, designed for bioconjugation and click chemistry applications.
  • Azide functionality enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Primary amine enables conjugation to activated carboxylates such as NHS esters.
  • Molecular weight approximately 614.73 g/mol; formula C26H54N4O12.
  • High purity by NMR (typically 98.0%).
  • Water-soluble PEG12 linker suitable for linker synthesis, PROTACs, and ADCs.
  • Store sealed at 4°C, protect from moisture and light.
Catalog No. 50-002-85004 Supplier Medchemexpress LLC Supplier No. HYW59056950MG
Azido-PEG12-amine is a PEG12 linker with an azide group at one terminus and a primary amine at the other, designed for bioconjugation and click chemistry applications.
  • Azide functionality enables copper-catalyzed azide-alkyne cycloaddition (click chemistry).
  • Primary amine enables conjugation to activated carboxylates such as NHS esters.
  • Molecular weight approximately 614.73 g/mol; formula C26H54N4O12.
  • High purity by NMR (typically 98.0%).
  • Water-soluble PEG12 linker suitable for linker synthesis, PROTACs, and ADCs.
  • Store sealed at 4°C, protect from moisture and light.