Learn More
Medchemexpress LLC Azido-PEG12-NHS ester | 2363756-50-5 | 98.0% | C31H56N4O16 | 50 MG
SDP
Catalog No. 5000353359
Encompass_Preferred
Azido-PEG12-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-53359 Supplier Medchemexpress LLC Supplier No. HY14076050MG
Azido-PEG12-NHS ester is a PEG-based PROTAC linker utilized in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups. Additionally, it can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Facilitates copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne-containing molecules
  • Can participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups