Learn More
Medchemexpress LLC Azido-PEG16-NHS ester | 00-00-0 | 99.9% | 917.00 | C39H72N4O20 | 5 MG
SDP
Catalog No. 5000222565
Encompass_Preferred
Azido-PEG16-NHS ester is a PEG-based N-hydroxysuccinimide (NHS) ester linker bearing an azide functional group for click chemistry and bioconjugation. It is used to introduce azide handles or to couple with primary amines when building conjugates, linkers, and PROTAC components in chemical biology workflows.
  • Contains an azide group for CuAAC and SPAAC click reactions.
  • NHS ester enables rapid coupling to primary amines.
  • PEG16 spacer provides hydrophilicity and conformational flexibility.
  • High purity suitable for analytical and preparative use.
  • Highly soluble in DMSO for concentrated stock solutions.
  • Stable when stored protected from light at 4°C; in solvent store at -80°C or -20°C.
Catalog No. 50-002-22565 Supplier Medchemexpress LLC Supplier No. HY1407615MG
Azido-PEG16-NHS ester is a PEG-based N-hydroxysuccinimide (NHS) ester linker bearing an azide functional group for click chemistry and bioconjugation. It is used to introduce azide handles or to couple with primary amines when building conjugates, linkers, and PROTAC components in chemical biology workflows.
  • Contains an azide group for CuAAC and SPAAC click reactions.
  • NHS ester enables rapid coupling to primary amines.
  • PEG16 spacer provides hydrophilicity and conformational flexibility.
  • High purity suitable for analytical and preparative use.
  • Highly soluble in DMSO for concentrated stock solutions.
  • Stable when stored protected from light at 4°C; in solvent store at -80°C or -20°C.