Learn More
Medchemexpress LLC Azido-PEG16-NHS ester | 00-00-0 | 99.9% | C39H72N4O20 | 10MG
SDP
Catalog No. 5000261404
Encompass_Preferred
Azido-PEG16-NHS ester is a PEG-based bifunctional linker featuring an azide moiety and an N-hydroxysuccinimide (NHS) ester. It enables conjugation to primary amines via the NHS ester and subsequent click chemistry using the azide, making it suitable for bioconjugation, labeling, and PROTAC synthesis. The reagent is provided at high purity and is soluble in DMSO for convenient formulation.
Catalog No. 50-002-61404 Supplier Medchemexpress LLC Supplier No. HY14076110MG
Azido-PEG16-NHS ester is a PEG-based bifunctional linker featuring an azide moiety and an N-hydroxysuccinimide (NHS) ester. It enables conjugation to primary amines via the NHS ester and subsequent click chemistry using the azide, making it suitable for bioconjugation, labeling, and PROTAC synthesis. The reagent is provided at high purity and is soluble in DMSO for convenient formulation.