Learn More
Medchemexpress LLC Azido-PEG2-azide | 59559-06-7 | 98.1% | 200.20 | 100 MG
SDP
Catalog No. 5000364696
Encompass_Preferred
Azido-PEG2-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Peg-based protac linker.
  • Used in the synthesis of protacs.
  • Click chemistry reagent containing an azide group.
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.
Catalog No. 50-003-64696 Supplier Medchemexpress LLC Supplier No. 100MGHYW044155
Azido-PEG2-azide is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. It functions as a click chemistry reagent due to its Azide group, enabling it to participate in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • Peg-based protac linker.
  • Used in the synthesis of protacs.
  • Click chemistry reagent containing an azide group.
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions.
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions.