Learn More
Medchemexpress LLC Azido-PEG2-C1-Boc | 251564-45-1 | >98% | C10H19N3O4 | 100 MG
SDP
Catalog No. 5000355699
Encompass_Preferred
Azido-PEG2-C1-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne group-containing molecules. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs, composed of two different ligands connected by a linker, leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-55699 Supplier Medchemexpress LLC Supplier No. HY140793100MG
Azido-PEG2-C1-Boc is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent containing an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) with Alkyne group-containing molecules. It also facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs, composed of two different ligands connected by a linker, leverage the intracellular ubiquitin-proteasome system to selectively degrade target proteins.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Facilitates strain-promoted alkyne-azide cycloaddition (SPAAC) reactions