Learn More
Medchemexpress LLC Azido-PEG2-CH2COOH | 882518-90-3 | 99.0% | 189.17 | 250 MG
SDP
Catalog No. 5000347341
Encompass_Preferred
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules containing DBCO or BCN groups.
  • PROTAC linker for PROTAC synthesis
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-47341 Supplier Medchemexpress LLC Supplier No. 250MGHY108368
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, containing an Azide group. This allows it to participate in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with molecules possessing Alkyne groups. Additionally, it can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions when combined with molecules containing DBCO or BCN groups.
  • PROTAC linker for PROTAC synthesis
  • Click chemistry reagent
  • Contains an Azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups