Learn More
Medchemexpress LLC Azido-PEG2-CH2COOH | 882518-90-3 | 99.03% | 189.17 | 1 ML
SDP
Catalog No. 5000346716
Encompass_Preferred
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs use the intracellular ubiquitin-proteasome system to degrade target proteins.
  • PEG-based PROTAC linker.
  • Click chemistry reagent.
  • Contains an Azide group for CuAAc reaction with Alkyne groups.
  • Can undergo SPAAC reactions with DBCO or BCN groups.
Catalog No. 50-003-46716 Supplier Medchemexpress LLC Supplier No. HY108368100MG
Add to Cart
Add to Cart
Azido-PEG2-CH2COOH is a PEG-based PROTAC linker used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group that can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups. PROTACs use the intracellular ubiquitin-proteasome system to degrade target proteins.
  • PEG-based PROTAC linker.
  • Click chemistry reagent.
  • Contains an Azide group for CuAAc reaction with Alkyne groups.
  • Can undergo SPAAC reactions with DBCO or BCN groups.