Learn More
Medchemexpress LLC Azido-PEG20-alcohol | 1637297-21-2 | >98% | C40H81N3O20 | 100 MG
SDP
Catalog No. 5000355700
Encompass_Preferred
Azido-PEG20-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-55700 Supplier Medchemexpress LLC Supplier No. HY140805100MG
Azido-PEG20-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is also a click chemistry reagent containing an Azide group, enabling copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with Alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in the synthesis of PROTACs
  • Click chemistry reagent
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups