Learn More
Medchemexpress LLC Azido-PEG20-alcohol | 1637297-21-2 | C40H81N3O20 | 25 MG
SDP
Catalog No. 5000352627
Encompass_Preferred
Azido-PEG20-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker for PROTAC synthesis.
  • Click chemistry reagent containing an Azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.
Catalog No. 50-003-52627 Supplier Medchemexpress LLC Supplier No. 25MGHY140805
Azido-PEG20-alcohol is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It is a click chemistry reagent containing an Azide group, capable of undergoing copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker for PROTAC synthesis.
  • Click chemistry reagent containing an Azide group.
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne groups.
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) with DBCO or BCN groups.