Learn More
Medchemexpress LLC Azido-PEG23-amine | 2172677-19-7 | 1023457 | 99.9% | C48H98N4O23 | 25 MG
SDP
Catalog No. 5000357387
Encompass_Preferred
Azido-PEG23-amine is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a versatile click chemistry reagent due to its azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with alkyne-containing molecules
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with DBCO or BCN group-containing molecules
Catalog No. 50-003-57387 Supplier Medchemexpress LLC Supplier No. 25MGHY140219
Azido-PEG23-amine is a PEG-based PROTAC linker primarily used in the synthesis of PROTACs. It functions as a versatile click chemistry reagent due to its azide group, enabling copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Used in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Reacts with alkyne-containing molecules
  • Enables strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Reacts with DBCO or BCN group-containing molecules