Learn More
Medchemexpress LLC Azido-PEG23-amine | 2172677-19-7 | 99.93% | C48H98N4O23 | 50 MG
SDP
Catalog No. 5000353010
Encompass_Preferred
Azido-PEG23-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing an azide group that enables various cycloaddition reactions.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reaction with alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
Catalog No. 50-003-53010 Supplier Medchemexpress LLC Supplier No. HY14021950MG
Azido-PEG23-amine is a PEG-based PROTAC linker that can be used in the synthesis of PROTACs. It functions as a click chemistry reagent, possessing an azide group that enables various cycloaddition reactions.
  • PEG-based PROTAC linker
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reaction with alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups