Learn More
Medchemexpress LLC Azido-PEG24-acid | 2195075-62-6 | 99.9% | C51H101N3O26 | 10MG
SDP
Catalog No. 5000260367
Encompass_Preferred
Azido-PEG24-acid is a polyethylene glycol (PEG)-based linker bearing a terminal azide, supplied as a solid for use in bioconjugation and linker synthesis. It facilitates efficient click chemistry conjugations and requires low-temperature, moisture-free storage to preserve stability.
  • PEG-based linker with terminal azide for click reactions.
  • Enables copper-catalyzed azide-alkyne cycloaddition and strain-promoted cycloaddition.
  • High purity (≈99.9%) suitable for synthesis and conjugation.
  • Molecular weight ≈1,172.35 g/mol for predictable spacer length.
  • Solid, white to light yellow appearance for easy handling.
  • Store sealed, away from moisture at low temperatures to maintain stability.
Catalog No. 50-002-60367 Supplier Medchemexpress LLC Supplier No. HY14045710MG
Azido-PEG24-acid is a polyethylene glycol (PEG)-based linker bearing a terminal azide, supplied as a solid for use in bioconjugation and linker synthesis. It facilitates efficient click chemistry conjugations and requires low-temperature, moisture-free storage to preserve stability.
  • PEG-based linker with terminal azide for click reactions.
  • Enables copper-catalyzed azide-alkyne cycloaddition and strain-promoted cycloaddition.
  • High purity (≈99.9%) suitable for synthesis and conjugation.
  • Molecular weight ≈1,172.35 g/mol for predictable spacer length.
  • Solid, white to light yellow appearance for easy handling.
  • Store sealed, away from moisture at low temperatures to maintain stability.