Learn More
Medchemexpress LLC Azido-PEG24-alcohol | 2415674-94-9 | 98.0% | C48H97N3O24 | 25 MG
SDP
Catalog No. 5000357096
Encompass_Preferred
Azido-PEG24-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Features an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Useful for cancer targeted therapy research
Catalog No. 50-003-57096 Supplier Medchemexpress LLC Supplier No. 25MGHY140806
Azido-PEG24-alcohol is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, featuring an Azide group that facilitates copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in PROTAC synthesis
  • Functions as a click chemistry reagent
  • Features an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAc) with alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups
  • Useful for cancer targeted therapy research