Learn More
Medchemexpress LLC Azido-PEG24-NHS ester | 2375600-46-5 | 95.0% | C55H104N4O28 | 10MG
SDP
Catalog No. 5000260177
Encompass_Preferred
Azido-PEG24-NHS ester is a polyethylene glycol (PEG) linker bearing an azide and an N-hydroxysuccinimide (NHS) ester functional group. It is designed for bioconjugation and linker synthesis, enabling both click chemistry and amine-directed coupling while improving solubility and flexibility in conjugates.
  • Azide functional group enables click chemistry with alkynes.
  • NHS ester reacts with primary amines to form stable amide bonds.
  • PEG24 spacer provides enhanced water solubility and flexible linker length.
  • Suitable for PROTAC synthesis, drug conjugates, and general bioconjugation.
  • High purity (~95.0%) suitable for research use.
  • Available in small milligram quantities for exploratory synthesis.
Catalog No. 50-002-60177 Supplier Medchemexpress LLC Supplier No. HY14076210MG
Azido-PEG24-NHS ester is a polyethylene glycol (PEG) linker bearing an azide and an N-hydroxysuccinimide (NHS) ester functional group. It is designed for bioconjugation and linker synthesis, enabling both click chemistry and amine-directed coupling while improving solubility and flexibility in conjugates.
  • Azide functional group enables click chemistry with alkynes.
  • NHS ester reacts with primary amines to form stable amide bonds.
  • PEG24 spacer provides enhanced water solubility and flexible linker length.
  • Suitable for PROTAC synthesis, drug conjugates, and general bioconjugation.
  • High purity (~95.0%) suitable for research use.
  • Available in small milligram quantities for exploratory synthesis.