Learn More
Medchemexpress LLC Azido-PEG3-aldehyde | 1807530-10-4 | MFCD28976706 | >95.0% | C9H17N3O4 | 10MG
SDP
Catalog No. 5000260998
Encompass_Preferred
Azido-PEG3-aldehyde is a PEG-based bifunctional linker bearing an azide and an aldehyde functional group. It is used to join molecules through click chemistry or aldehyde-based ligation reactions, enabling construction of linkers, probes, and conjugates for research applications. Molecular formula C9H17N3O4; molecular weight 231.25 g/mol.
  • Provides an azide for click chemistry conjugation.
  • Contains an aldehyde for reductive amination or oxime ligation.
  • Compatible with CuAAC and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Suitable for linker assembly, probe labeling, and bioconjugation workflows.
  • Available in small, research-scale quantities suitable for synthesis and labeling.
Catalog No. 50-002-60998 Supplier Medchemexpress LLC Supplier No. HY13851810MG
Azido-PEG3-aldehyde is a PEG-based bifunctional linker bearing an azide and an aldehyde functional group. It is used to join molecules through click chemistry or aldehyde-based ligation reactions, enabling construction of linkers, probes, and conjugates for research applications. Molecular formula C9H17N3O4; molecular weight 231.25 g/mol.
  • Provides an azide for click chemistry conjugation.
  • Contains an aldehyde for reductive amination or oxime ligation.
  • Compatible with CuAAC and strain-promoted alkyne-azide cycloaddition (SPAAC).
  • Suitable for linker assembly, probe labeling, and bioconjugation workflows.
  • Available in small, research-scale quantities suitable for synthesis and labeling.