Learn More
Medchemexpress LLC Azido-PEG3-C3-OH | 1807512-36-2 | 98.0% | C9H19N3O4 | 250 MG
SDP
Catalog No. 5000352603
Encompass_Preferred
Azido-PEG3-C3-OH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This click chemistry reagent features an Azide group, facilitating copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It also supports strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions
Catalog No. 50-003-52603 Supplier Medchemexpress LLC Supplier No. 250MGHY140808
Azido-PEG3-C3-OH is a PEG-based PROTAC linker designed for use in the synthesis of PROTACs. This click chemistry reagent features an Azide group, facilitating copper-catalyzed azide-alkyne cycloaddition (CuAAc) with alkyne-containing molecules. It also supports strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • PEG-based PROTAC linker
  • Utilized in the synthesis of PROTACs
  • Functions as a click chemistry reagent
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions