Learn More
Medchemexpress LLC Azido-PEG3-C6-Cl | 1625717-44-3 | MFCD34471373 | 95.3% | C12H24ClN3O3 | 10MG
SDP
Catalog No. 5000260559
Encompass_Preferred
Azido-PEG3-C6-Cl is a PEG-based PROTAC linker and click-chemistry reagent containing an azide functional group. It has molecular formula C12H24ClN3O3, molecular weight 293.79 g/mol, and reported purity 95.3%. The product is a colorless to light-yellow liquid supplied in small milligram pack sizes for research use and is stored at -20°C in pure form (stable up to 3 years).
  • Acts as a PEG-based linker for PROTAC and conjugate synthesis.
  • Contains an azide group compatible with CuAAC and strain-promoted click reactions.
  • Reported purity of 95.3% suitable for research applications.
  • Available in small pack sizes for synthetic and screening workflows.
  • Stable when stored at recommended temperatures, enabling longer-term storage.
Catalog No. 50-002-60559 Supplier Medchemexpress LLC Supplier No. HY13846610MG
Azido-PEG3-C6-Cl is a PEG-based PROTAC linker and click-chemistry reagent containing an azide functional group. It has molecular formula C12H24ClN3O3, molecular weight 293.79 g/mol, and reported purity 95.3%. The product is a colorless to light-yellow liquid supplied in small milligram pack sizes for research use and is stored at -20°C in pure form (stable up to 3 years).
  • Acts as a PEG-based linker for PROTAC and conjugate synthesis.
  • Contains an azide group compatible with CuAAC and strain-promoted click reactions.
  • Reported purity of 95.3% suitable for research applications.
  • Available in small pack sizes for synthetic and screening workflows.
  • Stable when stored at recommended temperatures, enabling longer-term storage.