Learn More
Azido-PEG3-carboxyrhodamine 110 conjugate, Thermo Scientific Chemicals
Description
It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is suitable for confocal laser scanning microscopy. Its long PEG spacer should reduce any steric effect of the dye on the molecule to be labeled. Carboxyrhodamine 110 and its derivatives are superior alternatives to other green fluorescent dyes.
Solubility
Soluble in DMSO, DMF or methanol.
Notes
Excite using the 488nm spectral line of the argon ion laser
Specifications
Specifications
| Color | Red |
| Quantity | 25 mg |
| Solubility Information | Soluble in DMSO,DMF or methanol. |
| Formula Weight | 574.59 |
| Physical Form | Solid |
| Chemical Name or Material | Azido-PEG3-carboxyrhodamine 110 conjugate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| TSCA | No |
| Recommended Storage | Store at -20°C |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.