Learn More
eMolecules​ 1092654-47-1 | Azido-PEG3-CH2CO2-NHS | Broadpharm | MFCD29067172 | 330.297 | C12H18N4O7 | 98.000 | [N-]=[N+]=NCCOCCOCCOCC(=O)ON1C(=O)CCC1=O | 500mg | 393649404
Catalog No. NC2000670 Shop All eMolecules Inc. Products
Encompass_Preferred
Azido-PEG3-CH2CO2-NHS | Broadpharm | 1092654-47-1 | MFCD29067172 | 330.297 | C12H18N4O7 | 98.000 | [N-]=[N+]=NCCOCCOCCOCC(=O)ON1C(=O)CCC1=O | 500mg | 393649404
Catalog No. NC2000670 Supplier eMolecules​ Supplier No. 393649404
Azido-PEG3-CH2CO2-NHS | Broadpharm | 1092654-47-1 | MFCD29067172 | 330.297 | C12H18N4O7 | 98.000 | [N-]=[N+]=NCCOCCOCCOCC(=O)ON1C(=O)CCC1=O | 500mg | 393649404