Learn More
Medchemexpress LLC Azido-PEG3-DYKDDDDK (Azido-PEG3-FLAG) | 2243206-95-1 | 1242.20 | 1 MG
SDP
Catalog No. 5000398402
Encompass_Preferred
Azido-PEG3-DYKDDDDK (Azido-PEG3-FLAG) is a multifunctional fusion tag designed for the purification of recombinant proteins. It functions as a click chemistry reagent, containing an Azide group.
  • Multifunctional fusion tag for the purification of recombinant proteins
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-98402 Supplier Medchemexpress LLC Supplier No. HYP5112A1MG
Azido-PEG3-DYKDDDDK (Azido-PEG3-FLAG) is a multifunctional fusion tag designed for the purification of recombinant proteins. It functions as a click chemistry reagent, containing an Azide group.
  • Multifunctional fusion tag for the purification of recombinant proteins
  • Click chemistry reagent
  • Contains an Azide group
  • Can undergo copper-catalyzed azide-alkyne cycloaddition reaction (CuAAc) with molecules containing alkyne groups
  • Can undergo strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups