Learn More
Medchemexpress LLC Azido-PEG3-DYKDDDDK (Azido-PEG3-FLAG) | 2243206-95-1 | 1242.20 | 10 MG
SDP
Catalog No. 5000397120
Encompass_Preferred
Azido-PEG3-DYKDDDDK (azide-PEG3-FLAG) is a multifunctional fusion tag for the purification of recombinant proteins. It is a click chemistry reagent that contains an azide group.
  • Incorporates an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAC) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups
Catalog No. 50-003-97120 Supplier Medchemexpress LLC Supplier No. HYP5112A10MG
Azido-PEG3-DYKDDDDK (azide-PEG3-FLAG) is a multifunctional fusion tag for the purification of recombinant proteins. It is a click chemistry reagent that contains an azide group.
  • Incorporates an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition reactions (CuAAC) with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules possessing DBCO or BCN groups