Learn More
Medchemexpress LLC Azido-peg3-dykddddk tfa | 1242.20 | 5 MG
SDP
Catalog No. 5000398253
Encompass_Preferred
Azido-PEG3-DYKDDDDK (azide-PEG3-FLAG) TFA is a multifunctional fusion tag used for the purification of recombinant proteins. This click chemistry reagent contains an azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Multifunctional fusion tag for the purification of recombinant proteins
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-98253 Supplier Medchemexpress LLC Supplier No. HYP5112A5MG
Azido-PEG3-DYKDDDDK (azide-PEG3-FLAG) TFA is a multifunctional fusion tag used for the purification of recombinant proteins. This click chemistry reagent contains an azide group, enabling it to undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups. It can also participate in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Multifunctional fusion tag for the purification of recombinant proteins
  • Contains an azide group
  • Enables copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups
  • Participates in strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups