Learn More
Medchemexpress LLC Azido-PEG3-DYKDDDDK (TFA) | 1242.20 | 50 MG
SDP
Catalog No. 5000398232
Encompass_Preferred
Azido-PEG3-DYKDDDDK (azide-PEG3-FLAG) TFA is a multifunctional fusion tag for the purification of recombinant proteins. It is a click chemistry reagent that contains an azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Multifunctional fusion tag for the purification of recombinant proteins
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reaction with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups
Catalog No. 50-003-98232 Supplier Medchemexpress LLC Supplier No. HYP5112A50MG
Azido-PEG3-DYKDDDDK (azide-PEG3-FLAG) TFA is a multifunctional fusion tag for the purification of recombinant proteins. It is a click chemistry reagent that contains an azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing alkyne groups, or strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Multifunctional fusion tag for the purification of recombinant proteins
  • Click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition (CuAAc) reaction with molecules containing alkyne groups
  • Undergoes strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups