Learn More
Medchemexpress LLC Azido-PEG3-DYKDDDDK TFA | 2243206-95-1 | 1242.20 | 25 MG
SDP
Catalog No. 5000400323
Encompass_Preferred
Azido-PEG3-DYKDDDDK TFA is a multifunctional fusion tag used for the purification of recombinant proteins. It functions as a click chemistry reagent, featuring an azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAC) with molecules containing alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Multifunctional fusion tag
  • Purifies recombinant proteins
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions
  • Participates in strain-promoted alkyne-azide cycloaddition reactions
Catalog No. 50-004-00323 Supplier Medchemexpress LLC Supplier No. 25MGHYP5112A
Azido-PEG3-DYKDDDDK TFA is a multifunctional fusion tag used for the purification of recombinant proteins. It functions as a click chemistry reagent, featuring an azide group. It can undergo copper-catalyzed azide-alkyne cycloaddition reactions (CuAAC) with molecules containing alkyne groups, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with molecules containing DBCO or BCN groups.
  • Multifunctional fusion tag
  • Purifies recombinant proteins
  • Functions as a click chemistry reagent
  • Contains an azide group
  • Undergoes copper-catalyzed azide-alkyne cycloaddition reactions
  • Participates in strain-promoted alkyne-azide cycloaddition reactions