Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Azido-PEG3-maleimide Kit

Catalog No. AAJ65984MC
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ65984MC 100 mg
AAJ65984MB 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAJ65984MC Supplier Thermo Scientific Chemicals Supplier No. J65984MC
  • $429.00
    $364.65 / Each of 1

    Save $64.35

    15% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart

missing translation for 'validMainframeMsg'

C15H23N5O6

An azido containing reagent with PEG spacer and sulfhydryl reactivity. The compound is not stable upon prolonged storage and is sold as a kit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An azido containing reagent with PEG spacer and sulfhydryl reactivity. The compound is not stable upon prolonged storage and is sold as a kit.

Solubility
Difficult to mix.

Notes
Kit contains two components: a) 1.1 eq. of azido-PEG3-amine as an oil; b) 1.0 eq. of maleimide-NHS ester as a solid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H23N5O6
MDL Number MFCD22380764
Synonym N-(11-Azido-3,6,9-trioxaundecyl)-3-(N-maleimidyl)propionamide

Specifications

Quantity 100 mg
UN Number UN2735
Solubility Information Difficult to mix.
Formula Weight 369.37
Chemical Name or Material Azido-PEG3-maleimide Kit
What storage conditions should I use for the Azido-PEG3-maleimide Kit?

The Azido-PEG3-maleimide Kit should be stored in the dark at -20°C.

What is the protocol for Azide-PEG3-maleimide generation with the Azido-PEG3-maleimide Kit?

We recommend the following protocol for Azide-PEG3-maleimide generation with the Azido-PEG3-maleimide Kit:

  1. For the 25 mg kit, add 1 mL of a dry water-miscible solvent (like DMF or DMSO) to the azido-PEG-amine and shake for approximately 30 seconds to ensure mixing. For the 100 mg kit add 2.5 mL of solvent.
  2. Place the maleimide-NHS ester under nitrogen or dry atmosphere and slowly add the solution from step 1 with stirring or shaking.
  3. Stir or shake the reaction mixture for 30 minutes at room temperature. The progress of the reaction can be followed by Thin Layer Chromatograph.
    Note: For TLC, the solvent is typically methanol:methylene chloride (1:20), run on a silica gel normal phase plate and developed with a potassium permanganate spray. The Rf of the azide-PEG3-maleimide is slightly lower than the Rf for the maleimide-NHS ester. When the reaction is complete, it will be one clean spot on the plate.
  4. The reaction product will be ready-to-use azide-PEG3-maleimide.
The azide-PEG3-maleimide is present at ~ 75 mM if 1 mL of solvent is used in step 1. The 25 mg kit produces 0.075 mmol of azide-PEG3-maleimide.

Hazard Category Warning
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
P261-P305+P351+P338-P302+P352-P321-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.