Learn More
Medchemexpress LLC Azido-PEG3-NHS ester | 1092654-47-1 | ≥98.0% | C12H18N4O7 | 250 MG
SDP
Catalog No. 5000357249
Encompass_Preferred
Azido-PEG3-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. This compound is integral in the development of PROTACs, which harness the ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables selective degradation of target proteins
Catalog No. 50-003-57249 Supplier Medchemexpress LLC Supplier No. 250MGHY140764
Azido-PEG3-NHS ester is a PEG-based PROTAC linker used in the synthesis of PROTACs. It functions as a click chemistry reagent, capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with alkyne-containing molecules, and strain-promoted alkyne-azide cycloaddition (SPAAC) reactions with DBCO or BCN groups. This compound is integral in the development of PROTACs, which harness the ubiquitin-proteasome system to selectively degrade target proteins.
  • Peg-based protac linker
  • Used in the synthesis of protacs
  • Click chemistry reagent
  • Contains an azide group
  • Participates in copper-catalyzed azide-alkyne cycloaddition (CuAAc)
  • Engages in strain-promoted alkyne-azide cycloaddition (SPAAC)
  • Enables selective degradation of target proteins