Learn More
Medchemexpress LLC Azido-PEG3-NHS ester | 1245718-89-1 | MFCD23726613 | 99.9% | 344.32 g/mol | C13H20N4O7 | 5 G
SDP
Catalog No. 5000202954
Encompass_Preferred
N3-PEG3-C2-NHS ester (Azido-PEG3-NHS ester) is a non-cleavable, azide-terminated three-unit polyethylene glycol linker bearing an N-hydroxysuccinimide (NHS) ester reactive group used for conjugation to primary amines and for click chemistry applications.
  • Non-cleavable azide-terminated PEG3 linker for bioconjugation.
  • NHS ester selectively reacts with primary amines for efficient coupling.
  • Compatible with azide-alkyne cycloaddition (click chemistry) for downstream functionalization.
  • High purity suitable for research applications.
  • Stable under recommended storage conditions: pure form at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-02954 Supplier Medchemexpress LLC Supplier No. HY1265285G
N3-PEG3-C2-NHS ester (Azido-PEG3-NHS ester) is a non-cleavable, azide-terminated three-unit polyethylene glycol linker bearing an N-hydroxysuccinimide (NHS) ester reactive group used for conjugation to primary amines and for click chemistry applications.
  • Non-cleavable azide-terminated PEG3 linker for bioconjugation.
  • NHS ester selectively reacts with primary amines for efficient coupling.
  • Compatible with azide-alkyne cycloaddition (click chemistry) for downstream functionalization.
  • High purity suitable for research applications.
  • Stable under recommended storage conditions: pure form at -20°C; in solvent at -80°C (6 months) or -20°C (1 month).